missing translation for 'onlineSavingsMsg'
Learn More

Ammonium persulfate, 98%, extra pure

Catalog No. p-3586394
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC201531000 100 g Glass bottle
AC201530010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC201531000 Supplier Thermo Scientific Chemicals Supplier No. 201531000
Only null left

CAS: 7727-54-0 | H8N2O8S2 | 228.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent

Chemical Identifiers

CAS 7727-54-0
Molecular Formula H8N2O8S2
Molecular Weight (g/mol) 228.19
MDL Number MFCD00003390
InChI Key ROOXNKNUYICQNP-UHFFFAOYSA-N
Synonym ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
PubChem CID 62648
IUPAC Name diazanium;sulfonatooxy sulfate
SMILES [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O

Specifications

Melting Point 120.0°C
Color White to Yellow
Physical Form Crystalline Powder and/or Crystals
Quantity 100 g
Assay Percent Range 98%
Linear Formula (NH4)2S2O8
Fieser 01,952; 02,348; 03,238; 05,15; 06,20; 12,33
Merck Index 15, 539
Solubility Information Solubility in water: 582g/L (20°C), may decompose.
Formula Weight 228.19
Percent Purity 98%
Grade Extra Pure
Packaging Glass bottle
Specific Gravity 1.98
Density 1.9800g/mL
Chemical Name or Material Ammonium persulfate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Take any precaution to avoid mixing with combustibles.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs.
EINECSNumber 231-786-5

RUO – Research Use Only