missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Peroxydisulfate, Purified, 98%, Spectrum™ Chemical

Catalog No. 18608752
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608752 50 kg Poly Pail
18608751 500 g Poly Pail
18608750 2.5 kg Poly Pail
18608749 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18608752 Supplier Spectrum Chemical Mfg Cor Supplier No. A123050KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7727-54-0

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 50 kg
Molecular Weight (g/mol) 228.19
Percent Purity 98%
Grade Purified
Packaging Poly Pail
Chemical Name or Material Ammonium Peroxydisulfate

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.