missing translation for 'onlineSavingsMsg'
Learn More

Ammonium peroxydisulfate, Electrophoresis Grade

Catalog No. p-7042251
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6185614 25 g
AAJ6185622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6185614 Supplier Thermo Scientific Chemicals Supplier No. J6185614
Only null left

CAS: 7727-54-0 | H8N2O8S2 | 228.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7727-54-0
Molecular Formula H8N2O8S2
Molecular Weight (g/mol) 228.19
MDL Number MFCD00003390
InChI Key ROOXNKNUYICQNP-UHFFFAOYSA-N
Synonym ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
PubChem CID 62648
IUPAC Name diazanium;sulfonatooxy sulfate
SMILES [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O

Specifications

Melting Point 120°C (decomposition)
Color White
Physical Form Powder
Quantity 25 g
Linear Formula (NH4)2S2O8
UN Number UN1444
Flash Point None
Merck Index 14,541
Formula Weight 228.2
Grade Electrophoresis
Odor Odorless
Refractive Index 1.5
Sensitivity Moisture sensitive
Density 1.982
Chemical Name or Material Ammonium peroxydisulfate
Show More Show Less
Hazard Category H272-H302-H315-H317-H319-H334-H335
Hazard Statement GHS H Statement
H272-H301-H334-H315-H319-H317-H335
May intensify fire; oxidizer.
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P330-P333+P313-P342+P311-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: AMMONIUM PERSULFATE
EINECSNumber 231-786-5
RTECSNumber SE0350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only