missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Peroxydisulfate, Crystal, BiotechGrade, 98%, Spectrum™ Chemical

Catalog No. p-7156920
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600636 100 g Poly Bottle
18600637 500 g Poly Bottle
18600638 2.5 kg Poly Bottle
18600639 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18600636 Supplier Spectrum Chemical Mfg Cor Supplier No. A1227100GM
Only null left

A1227, 7727-54-0, (NH4)2S2O8

Spectrum™ Chemical Ammonium Peroxydisulfate, Crystal, BiotechGrade this highly water soluble oxidizing agent is a catalyst for acrylamide gel polymerization when used in conjunction with Tetramethylethylenediamine. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.

Chemical Identifiers

CAS 7727-54-0
Molecular Formula H8N2O8S2
Molecular Weight (g/mol) 228.19
MDL Number MFCD00003390
InChI Key ROOXNKNUYICQNP-UHFFFAOYSA-N
IUPAC Name diammonium [(sulfonatoperoxy)sulfonyl]oxidanide
SMILES [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O

Specifications

CAS Max % 100%
Quantity 100 g
UN Number UN1444
Percent Purity 98%
Grade Biotech
Packaging Poly Bottle
Residue after Ignition 0.05%
Appearance Colorless to white