missing translation for 'onlineSavingsMsg'
Learn More

Ammonium peroxydisulfate, 98%

Catalog No. p-7080820
Change view
Click to view available options
Quantity:
500 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1053336 500 g
AAA105330B 1000 g
AAA105330I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1053336 Supplier Thermo Scientific Chemicals Supplier No. A1053336
Only null left

CAS: 7727-54-0 | H8N2O8S2 | 228.19 g/mol

Ammonium peroxydisulfate is widely utilized as an oxidizing agent and radical initiators in the polymerization of certain alkenes. It is actively involved in in the synthesis of commercially important polymers like styrene-butadiene rubber and polytetrafluoroethylene. It combines with tetramethylethylenediamine to catalyze the polymerization of acrylamide. Due to its oxidizing properties, it can be employed to etch copper on printed circuit boards which is an alternative to ferric chloride solution. Also, it acts as oxidants in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium peroxydisulfate is widely utilized as an oxidizing agent and radical initiators in the polymerization of certain alkenes. It is actively involved in in the synthesis of commercially important polymers like styrene-butadiene rubber and polytetrafluoroethylene. It combines with tetramethylethylenediamine to catalyze the polymerization of acrylamide. Due to its oxidizing properties, it can be employed to etch copper on printed circuit boards which is an alternative to ferric chloride solution. Also, it acts as oxidants in organic synthesis.

Solubility
Freely soluble in water.

Notes
Aqueous solution decomposes slowly at room temperature.Moisture sensitive. Incompatible with strong reducing agents, organic materials and powdered metals.

Chemical Identifiers

CAS 7727-54-0
Molecular Formula H8N2O8S2
Molecular Weight (g/mol) 228.19
MDL Number MFCD00003390
InChI Key ROOXNKNUYICQNP-UHFFFAOYSA-N
Synonym ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
PubChem CID 62648
IUPAC Name diazanium;sulfonatooxy sulfate
SMILES [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O

Specifications

Melting Point 120°C (decomposition)
Quantity 500 g
Linear Formula (NH4)2S2O8
UN Number UN1444
Flash Point None
Merck Index 14,541
Solubility Information Freely soluble in water.
Formula Weight 228.19
Percent Purity 98%
Odor Odorless
Refractive Index 1.5
Sensitivity Moisture sensitive
Density 1.982
Chemical Name or Material Ammonium peroxydisulfate
Show More Show Less
Hazard Category H272-H302-H315-H317-H319-H334-H335
Hazard Statement GHS H Statement
H272-H301-H334-H315-H319-H317-H335
May intensify fire
oxidizer.
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P330-P333+P313-P342+P311-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: AMMONIUM PERSULFATE
EINECSNumber 231-786-5
RTECSNumber SE0350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only