missing translation for 'onlineSavingsMsg'
Learn More

Ammonium pentaborate octahydrate, 99.0%

Catalog No. AA12294A3
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA12294A3 2 kg
AA1229436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA12294A3 Supplier Thermo Scientific Chemicals Supplier No. 012294A3
Only null left

CAS: 12046-03-6 | (NH4)2B10O16·8H2O

Ammonium pentaborate octahydrate is used as a component of electrolyte solutions in electrolytic capacitors. It is also used as a flame retarder and in wood and textile industry. Further, it is used for brazing, welding, soldering fluxes and in some detergents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12046-03-6
MDL Number MFCD00135839

Specifications

Physical Form Powder
Quantity 2 kg
Assay Percent Range 99.0%
Molecular Formula (NH4)2B10O16·8H2O
Solubility Information Soluble in water. Insoluble in ethanol.
Formula Weight 544.29 (400.17 Anhydrous)
Chemical Name or Material Ammonium pentaborate octahydrate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 234-521-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only