missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Oxalate Monohydrate, Honeywell Fluka™

Catalog No. p-7072065
Change view
Click to view available options
Quantity:
250 g
500 g
1 kg
Catalog No. Quantity
6002413 250 g
6002414 500 g
6002411 1 kg
3 options

Catalog No. 6002413

Supplier: Honeywell Chemicals 32304250G

Only null left

Puriss. p.a., ACS Reagent, Reagent ISO, Reagent European Pharmacopoeia, 99.5-101.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 6009-70-7
Molecular Formula C2H10N2O5
Molecular Weight (g/mol) 142.111
MDL Number MFCD00149694
InChI Key MSMNVXKYCPHLLN-UHFFFAOYSA-N
Synonym ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate
PubChem CID 516808
IUPAC Name diazanium;oxalate;hydrate
SMILES C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O

Specifications

Quantity 250 g
Linear Formula (NH4)2C2O4 · H2O
UN Number NONH for all modes of transport
Beilstein 3759830
Formula Weight 142.11g/mol
Percent Purity 99.5 to 101%
Grade ACS/puriss. p.a.
Residue after Ignition ≤0.02% (as SO4)
Chemical Name or Material Ammonium Oxalate Monohydrate, Reag. Ph. Eur. reag. ISO
Hazard Statement P301 + P312 + P330-P302 + P352 + P312
Precautionary Statement H302 + H312
EINECSNumber 238-135-4