missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Oxalate Monohydrate, greaterthan equal99%, ACS, MP Biomedicals

Catalog No. ICN19140880
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19140880 100 g
ICN19140890 500 g
ICN19140891 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19140880 Supplier MP Biomedicals Inc Supplier No. 0219140880

Chemical Identifiers

CAS 6009-70-7
Molecular Formula C2H10N2O5
Molecular Weight (g/mol) 142.111
MDL Number Number MFCD00149694
InChI Key MSMNVXKYCPHLLN-UHFFFAOYSA-N
Synonym ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate
PubChem CID 516808
IUPAC Name diazanium;oxalate;hydrate
SMILES C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O

Specifications

Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range ≥99%
Formula Weight 142.1
Grade ACS
Chemical Name or Material Ammonium Oxalate Monohydrate, Monohydrate
EINECSNumber 238-135-4