missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Oxalate Monohydrate (Crystalline/Certified ACS), Fisher Chemical

Supplier:  Thermo Fisher Scientific FLA679500

Catalog No. A679-500



Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ammonium Oxalate Monohydrate
6009-70-7
6.4
500 g
C2H10N2O5
MFCD00149694
ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate
C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O
142.111
142.11
Certified ACS
Poly Bottle
70°C
White
Solid
99.0 to 101.0%
(NH4)O(O)CC(O)O(NH4)·H2O
2811
MSMNVXKYCPHLLN-UHFFFAOYSA-N
diazanium;oxalate;hydrate
516808
99.0 to 101.0%
Pass Test
1.5g/mL

Chemical Identifiers

6009-70-7
142.111
MSMNVXKYCPHLLN-UHFFFAOYSA-N
516808
C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O
C2H10N2O5
MFCD00149694
ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate
diazanium;oxalate;hydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

DOTInformation : Class 6.1: Poison

SDS Documents

Documents

Promotions

Promotions