missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Oxalate, Monohydrate, Crystal, ACS, 99.0-101.0%, Spectrum™ Chemical

Catalog No. 18608731
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608731 50 kg Poly Bottle
18608726 25 g Poly Bottle
18608727 125 g Poly Bottle
18608728 500 g Poly Bottle
18608729 2.5 kg Poly Bottle
18608730 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18608731 Supplier Spectrum Chemical Mfg Cor Supplier No. A122050KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6009-70-7

Specifications

CAS Max % 1
Physical Form Crystals and/or Chunks
Quantity 50 kg
Molecular Weight (g/mol) 140.11
Percent Purity 99 to 101%
Grade Reagent
Packaging Poly Bottle
Residue after Ignition 0.0002
Chemical Name or Material Ammonium Oxalate, Monohydrate, Crystal

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.