missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium oxalate monohydrate, 99+%, ACS reagent
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 70.0°C |
| Color | White |
| Physical Form | Crystalline Powder |
| Quantity | 100 g |
| Packaging | Plastic bottle |
| Residue after Ignition | 0.02% max |
| Assay Percent Range | 99+% |
| Linear Formula | (CO2NH4)2·H2O |
| Merck Index | 15, 534 |
| Solubility Information | Solubility in water: 45g/L (20°C). Other solubilities: slightly soluble in alcohol |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Harmful in contact with skin. Harmful if swallowed. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Call a POISON CENTER or doctor/physician if you feel unwell. |
RUO – Research Use Only