Learn More
Ammonium O,O'-dimethyldithiophosphate, 95%
CAS: 1066-97-3 | C2H10NO2PS2 | 175.201 g/mol
$91.44 - $591.38
Chemical Identifiers
| CAS | 1066-97-3 |
|---|---|
| Molecular Formula | C2H10NO2PS2 |
| Molecular Weight (g/mol) | 175.201 |
| MDL Number | MFCD09753116 |
| InChI Key | PPGORMGERPBFTJ-UHFFFAOYSA-N |
| Synonym | ammonium o,o-dimethyl dithiophosphate, ammonium o,o'-dimethyldithiophosphate, ammonium o,o-dimethyl sulfanidylphosphonothioate, ammonium dimethyl phosphorodithioate, ammonium o,o'-dimethylphosphorodithioate, azanium dimethoxy-sulfanylidene-sulfido, o,o-dimethylphosphorodithioic acid ammonium salt, ammonium dimethoxy-sulfido-thioxo-$l^ 5-phosphane, azanium;dimethoxy-sulfanylidene-sulfido-?^ 5-phosphane |
| PubChem CID | 6451175 |
| IUPAC Name | azanium;dimethoxy-sulfanylidene-sulfido-$l^{5}-phosphane |
| SMILES | COP(=S)(OC)[S-].[NH4+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH3259203
|
Thermo Scientific Chemicals
H3259203 |
1 g |
Each for $91.44
|
|
|||||
|
AAH3259209
|
Thermo Scientific Chemicals
H3259209 |
10 g |
Each for $591.38
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1066-97-3 | |
| 175.201 | |
| PPGORMGERPBFTJ-UHFFFAOYSA-N | |
| 6451175 | |
| COP(=S)(OC)[S-].[NH4+] |
| C2H10NO2PS2 | |
| MFCD09753116 | |
| ammonium o,o-dimethyl dithiophosphate, ammonium o,o'-dimethyldithiophosphate, ammonium o,o-dimethyl sulfanidylphosphonothioate, ammonium dimethyl phosphorodithioate, ammonium o,o'-dimethylphosphorodithioate, azanium dimethoxy-sulfanylidene-sulfido, o,o-dimethylphosphorodithioic acid ammonium salt, ammonium dimethoxy-sulfido-thioxo-$l^ 5-phosphane, azanium;dimethoxy-sulfanylidene-sulfido-?^ 5-phosphane | |
| azanium;dimethoxy-sulfanylidene-sulfido-$l^{5}-phosphane |
Specifications
| 1066-97-3 | |
| Weak | |
| MFCD09753116 | |
| ammonium o,o-dimethyl dithiophosphate, ammonium o,o'-dimethyldithiophosphate, ammonium o,o-dimethyl sulfanidylphosphonothioate, ammonium dimethyl phosphorodithioate, ammonium o,o'-dimethylphosphorodithioate, azanium dimethoxy-sulfanylidene-sulfido, o,o-dimethylphosphorodithioic acid ammonium salt, ammonium dimethoxy-sulfido-thioxo-$l^ 5-phosphane, azanium;dimethoxy-sulfanylidene-sulfido-?^ 5-phosphane | |
| COP(=S)(OC)[S-].[NH4+] | |
| 175.201 | |
| 175.21 | |
| Ammonium O,O'-dimethyldithiophosphate |
| 146°C to 150°C | |
| C2H10NO2PS2 | |
| 1 g | |
| PPGORMGERPBFTJ-UHFFFAOYSA-N | |
| azanium;dimethoxy-sulfanylidene-sulfido-$l^{5}-phosphane | |
| 6451175 | |
| 95% |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
H302-H315-H319-H335
EINECSNumber : 213-922-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only