missing translation for 'onlineSavingsMsg'
Learn More

Ammonium O,O'-dimethyldithiophosphate, 95%

Catalog No. AAH3259209
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3259209 10 g
AAH3259203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3259209 Supplier Thermo Scientific Chemicals Supplier No. H3259209

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1066-97-3 | C2H10NO2PS2 | 175.201 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1066-97-3
Molecular Formula C2H10NO2PS2
Molecular Weight (g/mol) 175.201
MDL Number MFCD09753116
InChI Key PPGORMGERPBFTJ-UHFFFAOYSA-N
Synonym ammonium o,o-dimethyl dithiophosphate, ammonium o,o'-dimethyldithiophosphate, ammonium o,o-dimethyl sulfanidylphosphonothioate, ammonium dimethyl phosphorodithioate, ammonium o,o'-dimethylphosphorodithioate, azanium dimethoxy-sulfanylidene-sulfido, o,o-dimethylphosphorodithioic acid ammonium salt, ammonium dimethoxy-sulfido-thioxo-$l^ 5-phosphane, azanium;dimethoxy-sulfanylidene-sulfido-?^ 5-phosphane
PubChem CID 6451175
IUPAC Name azanium;dimethoxy-sulfanylidene-sulfido-$l^{5}-phosphane
SMILES COP(=S)(OC)[S-].[NH4+]

Specifications

Melting Point 146°C to 150°C
Odor Weak
Quantity 10 g
Formula Weight 175.21
Percent Purity 95%
Chemical Name or Material Ammonium O,O'-dimethyldithiophosphate
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
EINECSNumber 213-922-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only