missing translation for 'onlineSavingsMsg'
Learn More

Ammonium molybdate(VI) tetrahydrate, ACS reagent

Catalog No. p-45577
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC423310050 5 g Glass Bottle
AC423311000 100 g Plastic bottle
AC423315000 500 g Glass Bottle
AC423310025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC423310050 Supplier Thermo Scientific Chemicals Supplier No. 423310050
Only null left

CAS: 12054-85-2 | H24Mo7N6O24·4H2O | 1235.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent

Chemical Identifiers

CAS 12054-85-2
Molecular Formula H24Mo7N6O24·4H2O
Molecular Weight (g/mol) 1235.86
MDL Number MFCD00167059

Specifications

Melting Point 190°C
Color White
Physical Form Crystalline Powder
Quantity 5 g
Linear Formula (NH4)6Mo7O24·4H2O
Merck Index 15, 530
Solubility Information Solubility in water: 400g/L (20°C). Other solubilities: practically insoluble in alcohol
Formula Weight 1235.86
Percent Purity 81.0 to 83.0% (MoO3)
Grade ACS Reagent
Packaging Glass Bottle
Specific Gravity 2.49
Decomposition Information >150°C
Density 2.4900g/mL
Chemical Name or Material Ammonium molybdate(VI) tetrahydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only