missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Molybdate Tetrahydrate, puriss. p.a. ACS Reagent, Honeywell Fluka™

Catalog No. p-7071750
Change view
Click to view available options
Quantity:
100 g
25 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001880 100 g
6001882 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001880 Supplier Honeywell Chemicals Supplier No. 09880100G
Only null left

Puriss. p.a., ACS reagent,≥99.0% (T)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 12054-85-2
MDL Number MFCD00167059

Specifications

Quantity 100 g
Linear Formula (NH4)6Mo7O24 · 4H2O
UN Number NONH for all modes of transport
Formula Weight 1235.86g/mol
Percent Purity ≥99% (T)
Grade ACS/puriss. p.a.
Packaging Plastic Bottle
Density 2.498g/mL at 25°C
Chemical Name or Material Ammonium Molybdate Tetrahydrate
EINECSNumber 234-320-9
RTECSNumber QA5076000