missing translation for 'onlineSavingsMsg'
Learn More

Ammonium molybdate tetrahydrate, For ACS analysis, 81 to 83% MoO3, MP Biomedicals™

Catalog No. p-4404923
Change view
Click to view available options
Quantity:
100 g
500 g
Catalog No. Quantity
ICN15249780 100 g
ICN15249790 500 g
2 options

Catalog No. ICN15249780

Supplier: MP Biomedicals Inc 0215249780

Only null left

Chemical Identifiers

CAS 12054-85-2
Molecular Formula (NH4)6 Mo7O24·4H2O
Synonym Molybdic acid ammonium salt tetrahydrate

Specifications

Color White
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 81 to 83% MoO3
Formula Weight 1235.86
Density 2.87g/mL
Chemical Name or Material Ammonium molybdate tetrahydrate, For ACS analysis