Learn More
Ammonium Molybdate Tetrahydrate (Crystalline/Certified ACS), Fisher Chemical™
Supplier: Fisher Chemical FLA67410
Specifications
| Ammonium Molybdate Tetrahydrate | |
| White | |
| 5.0 to 5.5 | |
| 10 kg | |
| H24MO7N6O24 · 4 H2O | |
| MFCD00167059 | |
| 1235.86 | |
| Certified ACS | |
| Poly Pail | |
| 2.490g/cm³ |
| 190°C | |
| 12054-85-2 | |
| Solid | |
| 81.0 to 83.0 % | |
| (NH4)6Mo7O24·4H2O | |
| Ammonium Heptamolybdate Tetrahydrate | |
| 81.0 to 83.0% | |
| Pass Test | |
| >150°C |
Chemical Identifiers
| 12054-85-2 | |
| H24MO7N6O24 · 4 H2O | |
| MFCD00167059 | |
| Ammonium Heptamolybdate Tetrahydrate |
Safety and Handling
WARNING!
Emergency Overview
Harmful if swallowed. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Obtain medical attention. Do not induce vomiting. Do not induce vomiting. Call a physician or Poison Control Center immediately.
NFPA
Health:2
Flammability:1
Instability:0