missing translation for 'onlineSavingsMsg'
Learn More

Ammonium molybdate tetrahydrate, ACS Reagent, Honeywell Fluka™

Catalog No. p-7072292
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
6 x 500 g
Catalog No. Quantity
6003008 100 g
6003011 500 g
6003009 1 kg
6003012 6 x 500 g
4 options

Catalog No. 6003008

Supplier: Honeywell Chemicals A7302100G

Only null left

ACS Reagent, 81.0-83.0% MoO3O basis

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 12054-85-2
MDL Number MFCD00167059

Specifications

Quantity 100 g
Linear Formula (NH4)6Mo7O24 · 4H2O
UN Number NONH for all modes of transport
Formula Weight 1235.86g/mol
Grade ACS Reagent
Concentration or Composition (by Analyte or Components) 81 to 83% (MoO3 basis)
Density 2.498g/mL at 25°C
Chemical Name or Material Ammonium molybdate tetrahydrate
RTECSNumber QA5076000