missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium molybdate tetrahydrate, ACS Reagent, Honeywell Fluka™
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Quantity | 6 x 500 g |
| Linear Formula | (NH4)6Mo7O24 · 4H2O |
| UN Number | NONH for all modes of transport |
| Formula Weight | 1235.86g/mol |
| Grade | ACS Reagent |
| Concentration or Composition (by Analyte or Components) | 81 to 83% (MoO3 basis) |
| Density | 2.498g/mL at 25°C |
| Chemical Name or Material | Ammonium molybdate tetrahydrate |
Safety and Handling
| RTECSNumber | QA5076000 |