missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Molybdate, Tetrahydrate, ACS Grade, 81.0 to 83.0%, LabChem™

Catalog No. LC111951
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC111951 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC111951 Supplier LabChem Supplier No. LC111951

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ACS grade, suitable for laboratory and manufacturing procedures.

Chemical Identifiers

CAS 12054-85-2
Molecular Weight (g/mol) 1235.86

Specifications

Color White
CAS Max % 100
Physical Form Crystals
Quantity 500 g
Linear Formula (NH4)6Mo7O24·4H2O
Solubility Information Soluble in water
Formula Weight 1235.86
Percent Purity 81.0 to 83.0%
Grade ACS
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 2.276g/cm3
Decomposition Information Gaseous Ammonia
Density 2.276g/cm3
Assay as MoO3
Chemical Name or Material Ammonium Molybdate, Tetrahydrate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes skin irritation and serious eye irritation.
May cause respiratory irritation.
Harmful to aquatic life.
Precautionary Statement GHS P Statement
Avoid breathing dust.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature