missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Molybdate, 10% (w/v), pH adjusted, Ricca Chemical

Catalog No. 6701
Change view
Click to view available options
Package: Natural Poly; Size: 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
6701 Package: Natural Poly; Size: 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6701 Supplier Ricca Chemical Company Supplier No. RI6701

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 12054-85-2, 7732-18-5, 1310-73-2
CAS Min % 9.0, 85.0, 0.1
CAS Max % 11.0, 88.0, 0.9
Molecular Formula H6MoNO2
Synonym Molybdic acid ammonium salt tetrahydrate, Hexammonium heptamolybdate 4-hydrate
InChI Key XCOMAVCHADVANQ-UHFFFAOYSA-O
SMILES [NH4+].O.[O--].[Mo]
IUPAC Name ammonium hydrate molybdenum oxidandiide
Molecular Weight (g/mol) 148.00
PubChem CID 3084102  
Packaging Natural Poly Bottle
Treatment(s) pH adjusted
Solvent or Matrix Aqueous Solution
Quantity 4 L
Concentration or Composition Notes 10% (w/v)
Chemical Name or Material Ammonium Molybdate
Show More Show Less