missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Metavanadate, (White to light yellow powder),MP Biomedicals

Catalog No. ICN19462480
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19462480 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19462480 Supplier MP Biomedicals Inc Supplier No. 0219462480

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Common laboratory vanadate reagent suitable for use as a catalyst, analytical reagent, or in industrial applications like photography and ink production.

Chemical Identifiers

CAS 7803-55-6
Molecular Formula H4NO3V
Molecular Weight (g/mol) 116.98
MDL Number MFCD00011430
InChI Key YBVKNHXQSRDWAA-UHFFFAOYSA-M
Synonym ammonium metavanadate, ammonium vanadate v, ammonium monovanadate, unii-fl85px638g, vanadate vo31-, ammonium, vanadate vo31-, ammonium 1:1, ammonium vanadium oxide, ammoniummetavanadate, vanadic acid, hvo3 , ammonium salt, ammonium meta-vanadate
PubChem CID 516859
IUPAC Name vanadiumoylol amine
SMILES N.O[V](=O)=O

Specifications

Color White to Yellow
Physical Form Powder
Quantity 100 g
Formula Weight 117
Grade Cell Culture
Chemical Name or Material Ammonium Metavanadate
Recommended Storage Room Temperature