missing translation for 'onlineSavingsMsg'
Learn More

Ammonium magnesium phosphate hexahydrate, 98%

Catalog No. AAA1749736
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1749736 500 g
AAA1749722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1749736 Supplier Thermo Scientific Chemicals Supplier No. A1749736

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13478-16-5 | H16MgNO10P | 245.404 g/mol

It is used as pharmaceutical intermediae.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediae.

Solubility
Insoluble in water.

Notes
Store in a cool place. Ensure proper ventilation.

Chemical Identifiers

CAS 13478-16-5
Molecular Formula H16MgNO10P
Molecular Weight (g/mol) 245.404
MDL Number MFCD00150461
InChI Key CKMXBZGNNVIXHC-UHFFFAOYSA-L
Synonym unii-aw3ejl1462, magnesium ammonium phosphate hexahydrate, acmc-20alsk, magnesium 2+ ion ammonium hexahydrate phosphate, magnesium 2+ ammonium hexahydrate phosphate, 7785-21-9 h3po4-ammonium-magnesium 1:1:1
PubChem CID 10220511
IUPAC Name azanium;magnesium;phosphate;hexahydrate
SMILES [NH4+].O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[Mg+2]

Specifications

Quantity 500 g
Density 1.711
Solubility Information Insoluble in water.
Formula Weight 245.41 (137.32 Anhydrous)
Percent Purity 98%
Chemical Name or Material Ammonium magnesium phosphate hexahydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 232-075-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only