missing translation for 'onlineSavingsMsg'
Learn More

Ammonium L-(+)-tartrate, 98%

Catalog No. p-3256894
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1765818 50 g
AA1765830 250 g
AA17658A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1765818 Supplier Thermo Scientific Chemicals Supplier No. 01765818

CAS: 3164-29-2 | C4H12N2O6 | 184.15 g/mol

Ammonium L-(+)-tartrate finds application in the textile and pharmaceutical industries. It is also used to study protein structural analysis, biochemicals and reagents, biological buffers, chelators and proteomics. It acts as a nitrogen source in biological applications and cell culture media. Further, it is used in the study of crystal optimization research and preliminary diffraction data analysis of the Smad1 MH1 domain bonded to a palindromic SBE DNA element.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3164-29-2
Molecular Formula C4H12N2O6
Molecular Weight (g/mol) 184.15
MDL Number MFCD00013073
InChI Key NGPGDYLVALNKEG-OLXYHTOASA-N
Synonym ammonium d-tartrate, diammonium l-+-tartrate, l-tartaric acid, ammonium salt, unii-ji8ud88354, diammonium l-tartrate, r-2,3-dihydroxybutanedioic acid diammonium salt, ammonium tartrate dibasic, butanedioic acid, 2,3-dihydroxy-2r,3r-, diammonium salt, ammonium l-+-tartrate, diammonium 2r,3r-2,3-dihydroxysuccinate
PubChem CID 13652278
ChEBI CHEBI:63075
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid diamine
SMILES N.N.O[C@H]([C@@H](O)C(O)=O)C(O)=O

Specifications

Color White
Physical Form Crystalline
Quantity 50 g
Beilstein 6120352
Solubility Information Soluble in water and alcohol.
Formula Weight 184.15
Percent Purity 98%
Odor Odorless
Sensitivity Hygroscopic
Density 1.6
Chemical Name or Material Ammonium L-(+)-tartrate
Show More Show Less
EINECSNumber 221-618-9
RTECSNumber WW8050000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only