missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Iron(III) Sulfate Solution Volumetric, 0.1 M (NH4)Fe(SO4)2, Solstice
Description
Solstice's high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Specifications
| Physical Form | Liquid |
| Quantity | 1 L |
| UN Number | UN3264 |
| Formula Weight | 266.01g/mol |
| Grade | Volumetric |
| Packaging | Poly Bottle |
| Chemical Name or Material | Ammonium iron(III) sulfate solution |