missing translation for 'onlineSavingsMsg'
Learn More

Ammonium iron(II) sulfate hydrate, Puratronic™, 99.999% (metals basis)

Catalog No. AA4075114
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4075114 25 g
AA4075103 1 g
AA4075106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4075114 Supplier Thermo Scientific Chemicals Supplier No. 04075114

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7783-85-9 | FeH20N2O14S2 | 392.13 g/mol

Ammonium iron(II) sulfate hydrate is widely utilized in analytical chemistry for titration purposes since it is affected lesser by oxygen in the air than iron(II) sulpahte. It is employed to measure high doses of gamma rays by using Fricke's dosemeter. It is involved in a variety of other applications from nanomaterials to general redox reactions. It is used in the medical field as an antianemic agent for treatment of iron deficiency.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7783-85-9
Molecular Formula FeH20N2O14S2
Molecular Weight (g/mol) 392.13
MDL Number MFCD00150530
InChI Key MQLVWQSVRZVNIP-UHFFFAOYSA-L
Synonym Iron(II) ammonium sulfate; Ferrous ammonium sulfate
IUPAC Name λ²-iron(2+) diammonium hexahydrate disulfate
SMILES [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Melting Point ∼100°C (decomposition)
Color Blue-Green
Physical Form Coarse Powder
Quantity 25 g
Odor Odorless
Sensitivity Air and light sensitive
Density 1.864 g/mL
Linear Formula (NH4)2Fe(SO4)2·xH2O (x≈6)
Merck Index 14,521
Solubility Information Soluble in water. Insoluble in alcohol.
Formula Weight 284.04 (Anhydrous)
Percent Purity 99.999%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Ammonium iron(II) sulfate hydrate
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 233-382-4
RTECSNumber BR6500000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only