Learn More
Ammonium iron(II) sulfate hexahydrate, ACS, 98.5-101.5%
Description
Polymerization catalyst, photographyAmmonium iron(II) sulfate hexahydrate is widely utilized in analytical chemistry for titration purposes since it is affected lesser by oxygen in the air than iron(II) sulpahte. It is employed to measure high doses of gamma rays by using Fricke's dosemeter. It is involved in a variety of other applications from nanomaterials to general redox reactions. It is used in the medical field as an antianemic agent for treatment of iron deficiency.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | ∽100°C (decomposition) |
| Color | Blue-Green |
| Physical Form | Crystalline |
| Quantity | 500 g |
| Assay Percent Range | 98.5-101.5% |
| Solubility Information | Soluble in water. Insoluble in alcohol. |
| Formula Weight | 392.13 (284.04 Anhydrous) |
| Grade | ACS |
| Odor | Odorless |
| Sensitivity | Air and light sensitive |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 233-382-4 |
| RTECSNumber | BR6500000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only