missing translation for 'onlineSavingsMsg'
Learn More

Ammonium iron(II) sulfate hexahydrate, 99%, ACS reagent

Catalog No. p-3734801
Change view
Click to view available options
Quantity:
100 g
500 g
3 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC423721000 100 g Plastic bottle
AC423725000 500 g Plastic bottle
AC423720030 3 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC423721000 Supplier Thermo Scientific Chemicals Supplier No. 423721000
Only null left

CAS: 7783-85-9 | FeH20N2O14S2 | 392.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7783-85-9
Molecular Formula FeH20N2O14S2
Molecular Weight (g/mol) 392.13
MDL Number MFCD00150530
InChI Key MQLVWQSVRZVNIP-UHFFFAOYSA-L
Synonym Ferrous ammonium sulfate, Mohr's salt
IUPAC Name λ²-iron(2+) diammonium hexahydrate disulfate
SMILES [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Melting Point 100.0°C
Color Blue-Green
Physical Form Crystals
Quantity 100 g
Assay Percent Range 99%
Linear Formula (NH4)2Fe(SO4)2·6H2O
Merck Index 15, 518
Solubility Information Solubility in water: 269g/L (20°C). Other solubilities: practically insoluble in alcohol
Formula Weight 392.14
Percent Purity 99%
Grade ACS Reagent
Packaging Plastic bottle
Chemical Name or Material Ammonium iron(II) sulfate hexahydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Recommended Storage due to inherent oxidation, ferric ion (Fe3+) content may increase on storage

RUO – Research Use Only