missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide (TraceMetal™ Grade), Fisher Chemical™

Catalog No. p-212713
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
HDPE Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A512P500 500 mL HDPE Bottle
A5124 4 L HDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A512P500 Supplier Seastar Chemicals Inc Supplier No. S010701TFIQ03

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name amine hydrate
SMILES N.O

Specifications

Color Colorless
Linear Formula NH4OH
Density 0.920g/cm³
Quantity 500 mL
Formula Weight 35.05
Grade TraceMetal
Packaging HDPE Bottle
Vapor Pressure 504 hPa at 50°C
Physical Form Liquid
Chemical Name or Material Ammonium Hydroxide
DOTInformation DOT Class 8, : Corrosive