Learn More
Ammonium hydroxide, extra pure, 10 wt% in water
CAS: 1336-21-6 | H5NO | 35.05 g/mol
Supplier: Thermo Scientific Chemicals 458680025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Ammonium hydroxide | |
| H5NO | |
| MFCD00066650 | |
| ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
| VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
| amine hydrate | |
| 14923 | |
| 35.05 | |
| 0.9 |
| 1336-21-6 | |
| NH4OH | |
| 0.9000g/mL | |
| 2.5 L | |
| N.O | |
| 35.05 | |
| CHEBI:18219 | |
| Extra Pure |
Chemical Identifiers
| 1336-21-6 | |
| 35.05 | |
| VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
| 14923 | |
| amine hydrate |
| H5NO | |
| MFCD00066650 | |
| ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
| CHEBI:18219 | |
| N.O |
Safety and Handling
GHS H Statement Causes severe skin burns and eye damage. May cause respiratory irritation. Harmful to aquatic life with long lasting effects.
GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Avoid release to the environment.
Danger
RUO – Research Use Only