missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hydroxide solution, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., ∼25% NH3 basis, Honeywell Fluka™

Catalog No. p-7071970
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
4 x 2.5 L
4 x 5 L
6 x 1 L
Packaging:
Glass Bottle
Plastic Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6000320 1 L Plastic Bottle
6000322 2.5 L Plastic Bottle
6000324 5 L Plastic Bottle
60045827 6 x 1 L Glass Bottle
60045826 6 x 1 L Plastic Bottle
60045823 4 x 2.5 L Plastic Bottle
60045825 4 x 5 L Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 6000320 Supplier Honeywell Chemicals Supplier No. 305011L
Only null left

Puriss. p.a., Reag. ISO, Reag. Ph. Eur., ∼25% NH3 basis

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name azanium;hydroxide
SMILES N.O

Specifications

Linear Formula NH4OH
Concentration 25% NH3 basis
UN Number 2672
Beilstein 3587154
Density 0.9g/mL at 25°C, 0.892 to 0.910g/mL at 20°C
Quantity 1 L
Explosion Limit 0.27
Formula Weight 35.05g/mol
Grade puriss. p.a.
Packaging Plastic Bottle
Chemical Name or Material Ammonium Hydroxide Solution, Reag. Ph. Eur. reag. ISO
Show More Show Less
Hazard Statement P260-P280-P301 + P312 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
Precautionary Statement H302-H314-H335-H410