missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hydroxide solution, Puriss., meets analytical specification of Ph. Eur., 25-30% NH3 basis, Solstice

Catalog No. 6000315
Change view
Click to view available options
Quantity:
1 L
2.5 L
25 kg
4 x 2.5 L
Packaging:
Plastic Bottle
Plastic Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6000315 1 L Plastic Bottle
6000316 2.5 L Plastic Bottle
60044563 4 x 2.5 L Plastic Bottle
6000318 25 kg Plastic Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 6000315 Supplier Solstice Supplier No. 050031L

Puriss., meets analytical specification of Ph. Eur., 25-30% NH3 basis

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
SMILES N.O

Specifications

Linear Formula NH4OH
Concentration 25 to 30% NH3 basis
UN Number 2672
Beilstein 3587154
Density 0.892 to 0.910g/mL at 20°C, 0.9g/mL at 25°C
Quantity 1 L
IUPAC Name azanium;hydroxide
Explosion Limit 0.27
Formula Weight 35.05g/mol
Grade puriss.
Packaging Plastic Bottle
Chemical Name or Material Ammonium Hydroxide Solution, Meets analytical specification of Ph.Eur.
Show More Show Less
Hazard Statement P273-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P391
Precautionary Statement H302-H314-H335-H400