missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium hydroxide solution, Puriss., 30-33% NH3 in H2O, Honeywell Fluka™
Puriss., 30-33% NH3 in H2O
Supplier: Honeywell-Fluka 0500225L
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
| Ammonium Hydroxide Solution | |
| H5NO | |
| MFCD00066650 | |
| 2672 | |
| 0.9g/mL at 25°C | |
| 2.5 L | |
| N.O | |
| 0.27 | |
| 14923 | |
| 35.05g/mol | |
| Glass Bottle |
| 1336-21-6 | |
| NH4OH | |
| 30 to 33% NH3 in H2O | |
| 3587154 | |
| ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
| VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
| azanium;hydroxide | |
| 35.05 | |
| CHEBI:18219 | |
| puriss. |
Chemical Identifiers
| 1336-21-6 | |
| 35.05 | |
| VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
| 14923 | |
| N.O |
| H5NO | |
| MFCD00066650 | |
| ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
| CHEBI:18219 |
Safety and Handling
P260-P280-P301 + P312 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
H302-H314-H335-H410