missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Hydroxide Solution, ACS Reagent, 28.0 to 30.0% NH3 basis, Solstice
Description
- Leading the Industry For Over 65 Years
- Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Linear Formula | NH4OH |
| Concentration | 28 to 30% NH3 basis |
| Beilstein | 3587154 |
| Density | 0.9g/mL at 25°C |
| Quantity | 500 mL |
| IUPAC Name | azanium;hydroxide |
| Explosion Limit | 0.27 |
| Formula Weight | 35.05g/mol |
| Grade | ACS Reagent |
| Packaging | Glass Bottle |
| Show More |