missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Hydroxide Solution, 5.0M, Honeywell Fluka™
Volumetric, 5.0Â M NH4OH (5.0N)
Supplier: Honeywell Chemicals 3186122L
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
| Ammonium Hydroxide Solution | |
| H5NO | |
| MFCD00066650 | |
| 3587154 | |
| ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
| VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
| amine hydrate | |
| 35.05 | |
| CHEBI:18219 |
| 1336-21-6 | |
| NH4OH | |
| 2672 | |
| 0.9g/mL at 25°C, 0.965g/mL at 25°C | |
| 2 L | |
| N.O | |
| 0.27 | |
| 14923 | |
| 35.05g/mol |
Chemical Identifiers
| 1336-21-6 | |
| 35.05 | |
| VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
| 14923 | |
| N.O |
| H5NO | |
| MFCD00066650 | |
| ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
| CHEBI:18219 |
Safety and Handling
P260-P280-P301 + P312 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
H302-H314-H335-H412