missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide (Optima™), Fisher Chemical™

Catalog No. p-212711
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A470250 250 mL
A470500 500 mL
A4701 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A470250 Supplier Seastar Chemicals Inc Supplier No. S020701TFIJ01
Only null left

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name amine hydrate
SMILES N.O

Specifications

Color Colorless
Linear Formula NH4OH
Merck Index 15, 491
Density 0.920g/cm³
Quantity 250 mL
Formula Weight 35.05
Grade OPTIMA
Packaging HDPE Bottle
Vapor Pressure 502 hPa at 50°C
Physical Form Liquid
Chemical Name or Material Ammonium Hydroxide
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
DOTInformation DOT Class 8, : Corrosive