missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Hydroxide (Certified ACS Plus), Fisher Chemical™
Supplier: Thermo Fisher Scientific A669C212
Specifications
| 12 | |
| Colorless | |
| 38°C | |
| -57°C | |
| H5NO | |
| MFCD00066650 | |
| 6 x 2.5 L | |
| N.O | |
| 35.05 | |
| CHEBI:18219 | |
| 28.0 to 30.0% (w/w) | |
| Pass test | |
| Glass Bottle | |
| 500 hPa at 20°C |
| Ammonium Hydroxide | |
| about 14.8N | |
| 1336-21-6,7664-41-7,7732-18-5 | |
| Assay: 28.0 to 30.0 w/w % | |
| NH4OH | |
| ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
| VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
| amine hydrate | |
| 14923 | |
| 35.05 | |
| Certified ACS Plus | |
| about 14.8N | |
| 3ppm max. | |
| Liquid |
Chemical Identifiers
| 1336-21-6 | |
| 35.05 | |
| VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
| 14923 | |
| amine hydrate |
| H5NO | |
| MFCD00066650 | |
| ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
| CHEBI:18219 | |
| N.O |
Safety and Handling
DOTInformation : DOT Class 8: Corrosive