missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide ACS, MilliporeSigma™

Catalog No. MAX13036
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
Poly Coated Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MAX13036 500 mL Glass Bottle
MAX1303P5 500 mL Poly Coated Glass Bottle
MAX13033 2.5 L Glass Bottle
23749901 2.5 L Poly Coated Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MAX13036 Supplier Technidata Supplier No. AX13036

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name amine hydrate
SMILES N.O

Specifications

Assay Percent Range 28 to 30%
Linear Formula NH4OH [NH3 in water]
Density 0.90g/cm3 (20°C)
Quantity 500 mL
Explosion Limit 15.4 to 33.6%(V)
Formula Weight 35.05
Grade ACS/EP/Reagent
Packaging Glass Bottle
Vapor Pressure 635 hPa (20°C)
Chemical Name or Material Ammonium Hydroxide, EMSURE™, For Analysis
Recommended Storage Store below +25°C