missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Hydroxide ACS, MilliporeSigma™
Specifications
Specifications
| Assay Percent Range | 28 to 30% |
| Linear Formula | NH4OH [NH3 in water] |
| Density | 0.90g/cm3 (20°C) |
| Quantity | 2.5 L |
| Explosion Limit | 15.4 to 33.6%(V) |
| Formula Weight | 35.05 |
| Grade | ACS/EP/Reagent |
| Packaging | Glass Bottle |
| Vapor Pressure | 635 hPa (20°C) |
| Chemical Name or Material | Ammonium Hydroxide, EMSURE™, For Analysis |
Safety and Handling
| Recommended Storage | Store below +25°C |