missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hydroxide, ACS, 28.0-30.0% NH{3}

Catalog No. AA3328530
Change view
Click to view available options
Quantity:
250 g
1 kg
5 x 1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3328530 250 g
AA33285A1 1 kg
AA33285D9 5 x 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3328530 Supplier Thermo Scientific Chemicals Supplier No. 03328530

CAS: 1336-21-6 | H5NO | 35.05 g/mol

Ammonium hydroxide is widely utilized as a leavening agent or acidity regulator in food production. It serves as a precursor to some alkyl amines and is also used in the tobacco industry for flavor enhancement and as a processing aid. During furniture making, it combines with tannic acid and is used to darken or stain wood by making it iron salts. In chemical laboratories, it used for qualitative inorganic analysis, as a complexant and as a base. It is used to clean gold, silve, and platinum jewelry. It is an active component of Tollens' reagent (consisting of a solution of silver nitrate and ammonia) and is used to determine the presence of aldehyde or alpha-hydroxy ketone functional groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
SMILES N.O

Specifications

Color Colorless
Assay Percent Range 28.0-30.0% NH3
Density 0.90 g/cm3
Solubility Information Miscible with water.
Quantity 250 g
IUPAC Name amine hydrate
Formula Weight 35.05
Grade ACS
Odor Ammonia-like
Physical Form Liquid
Chemical Name or Material Ammonium hydroxide
Show More Show Less
Hazard Category H314-H335
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation DOT Class: 8, Packaging Group: III
EINECSNumber 215-647-6
RTECSNumber BQ9625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only