missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Hydroxide, 5% (v/v), Ricca Chemical
Specifications
Specifications
| Quantity | 20 L |
| Percent Purity | 4.9 to 5.1% |
| Grade | Laboratory |
| Packaging | Cubitainer |
| Chemical Name or Material | Ammonium Hydroxide Solution |