Learn More
Ammonium hydroxide, 5.0N Standardized Solution
CAS: 1336-21-6 | H5NO | 35.05 g/mol
$163.82 - $285.99
Chemical Identifiers
| CAS | 1336-21-6 |
|---|---|
| Molecular Formula | H5NO |
| Molecular Weight (g/mol) | 35.05 |
| MDL Number | MFCD00066650 |
| InChI Key | VHUUQVKOLVNVRT-UHFFFAOYSA-N |
| Synonym | ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh |
| PubChem CID | 14923 |
| ChEBI | CHEBI:18219 |
| IUPAC Name | amine hydrate |
| SMILES | N.O |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1336-21-6 | |
| 35.05 | |
| VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
| 14923 | |
| amine hydrate |
| H5NO | |
| MFCD00066650 | |
| ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
| CHEBI:18219 | |
| N.O |
Specifications
| Colorless | |
| Liquid | |
| H5NO | |
| ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
| N.O | |
| 35.05 | |
| CHEBI:18219 | |
| Colorless liquid |
| 1336-21-6 | |
| 1 L | |
| MFCD00066650 | |
| VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
| amine hydrate | |
| 14923 | |
| 35.05 | |
| Ammonium hydroxide |
Safety and Handling
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : DOT Class: 8; Packing Group: III
EINECSNumber : 215-647-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only