missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide 29% LM Prime, Transene

Catalog No. 502744717
Encompass
Change view
Click to view available options
Quantity:
1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
502744717 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 502744717 Supplier Doe and Ingalls Management LLC Supplier No. T0701818GAL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ammonium Hydroxide, 29 Percent, Electronic/Cleanroom Grade

This is a highly purified solution of ammonia in water, containing 29% ammonia by weight, specifically designed for use in the electronics industry due to its stringent purity standards, often used in semiconductor manufacturing processes like wafer cleaning and etching; also known as 'cleanroom grade' ammonium hydroxide.

Chemical Identifiers

CAS 1336-21-6
Molecular Weight (g/mol) 35 g/mol

Specifications

Quantity 1 gal.
Percent Purity 30%
Grade Electronic
Packaging HDPE
Chemical Name or Material Ammonium Hydroxide