missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Hydroxide, 16.7% (v/v), Electronic Grade, Ricca Chemical
Supplier: Ricca Chemical Company 6321
Specifications
| Ammonium Hydroxide | |
| 1336-21-6,7732-18-5 | |
| 84.0,17.0 | |
| MFCD00066650 | |
| ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
| VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
| amine hydrate | |
| 14923 | |
| Electronic | |
| Aqueous Solution |
| Colorless | |
| 83.0,16.0 | |
| H5NO | |
| 16.7% (v/v) | |
| 4 L | |
| N.O | |
| 35.05 | |
| CHEBI:18219 | |
| Natural Poly Bottle | |
| Liquid |
Chemical Identifiers
| 1336-21-6 | |
| 35.05 | |
| VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
| 14923 | |
| amine hydrate |
| H5NO | |
| MFCD00066650 | |
| ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
| CHEBI:18219 | |
| N.O |