missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hydrogen fluoride, tech. 95%, Thermo Scientific Chemicals

Catalog No. p-7049549
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1466036 500 g
AA14660A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1466036 Supplier Thermo Scientific Chemicals Supplier No. 01466036
Only null left

Ammonium hydrogen fluoride is used as a chemical reagent in analysis and an antiseptic. It is also used in ceramics, electroplating, component of glass etchants as well as food processing equipment disinfectant. It acts as an intermediate in the production of hydrofluoric acid from hexafluorosilicic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1341-49-7
Molecular Formula F2H5N
Molecular Weight (g/mol) 57.04
MDL Number MFCD00012018
InChI Key KVBCYCWRDBDGBG-UHFFFAOYSA-N
Synonym Ammonium bifluoride
IUPAC Name amine dihydrofluoride
SMILES N.F.F

Specifications

Boiling Point 238°C
Melting Point 120°C to 122°C
pH 2 to 3
Physical Form Flakes
Quantity 500 g
UN Number UN1727
Merck Index 14,498
Solubility Information Soluble in water. Slightly soluble in alcohol.
Formula Weight 57.04
Percent Purity 95%
Grade Technical
Odor Ammonia-like
Sensitivity Hygroscopic
Density 1.500 g/mL
Chemical Name or Material Ammonium hydrogen fluoride
Show More Show Less
Hazard Category H301-H314-H335
Hazard Statement GHS H Statement
H300-H314-H318
Fatal if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMMONIUM HYDROGENDIFLUORIDE, SOLID
EINECSNumber 215-676-4
RTECSNumber BQ9200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only