missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hydrogen fluoride, 95%, remainder mainly ammonium fluoride, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L172370E

 View more versions of this product

Catalog No. AAL172370E


Only null left
Explore available promotions

Description

Description

Employed in a new hydrothermal synthesis of complex fluorides NaHoF4 and NaEuF4 these interesting compounds have the fluorite structure and are important for solid-state lasers as well as for photoluminescence and magnetic characteristics

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed in a new hydrothermal synthesis of complex fluorides NaHoF4 and NaEuF4 these interesting compounds have the fluorite structure and are important for solid-state lasers as well as for photoluminescence and magnetic characteristics

Solubility
Soluble in water.

Notes
Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

238°C (decomposition)
120°C to 122°C
2 to 3
remainder mainly ammonium fluoride
(NH4)HF2
UN1727
Ammonium bifluoride
KVBCYCWRDBDGBG-UHFFFAOYSA-N
amine dihydrofluoride
57.04
Ammonia-like
1.5
Ammonium hydrogen fluoride
1341-49-7
2500 g
F2H5N
MFCD00012018
14,498
Soluble in water.
N.F.F
57.04
95%
Hygroscopic

Chemical Identifiers

1341-49-7
57.04
KVBCYCWRDBDGBG-UHFFFAOYSA-N
amine dihydrofluoride
F2H5N
MFCD00012018
Ammonium bifluoride
N.F.F
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H314-H318
Fatal if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H301-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMMONIUM HYDROGENDIFLUORIDE, SOLID

EINECSNumber : 215-676-4

RTECSNumber : BQ9200000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only