Learn More
Ammonium hydrogen fluoride, 95%, remainder mainly ammonium fluoride, Thermo Scientific Chemicals
Description
Employed in a new hydrothermal synthesis of complex fluorides NaHoF4 and NaEuF4 these interesting compounds have the fluorite structure and are important for solid-state lasers as well as for photoluminescence and magnetic characteristics
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Employed in a new hydrothermal synthesis of complex fluorides NaHoF4 and NaEuF4 these interesting compounds have the fluorite structure and are important for solid-state lasers as well as for photoluminescence and magnetic characteristics
Solubility
Soluble in water.
Notes
Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Boiling Point | 238°C (decomposition) |
| Melting Point | 120°C to 122°C |
| pH | 2 to 3 |
| Quantity | 500 g |
| Odor | Ammonia-like |
| Sensitivity | Hygroscopic |
| Density | 1.5 |
| Linear Formula | (NH4)HF2 |
| UN Number | UN1727 |
| Merck Index | 14,498 |
| Show More |
Safety and Handling
| Hazard Category | H301-H314-H335 |
| Hazard Statement | GHS H Statement H300-H314-H318 Fatal if swallowed. Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMMONIUM HYDROGENDIFLUORIDE, SOLID |
| EINECSNumber | 215-676-4 |
| RTECSNumber | BQ9200000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only