missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hydrogen fluoride, 95%, remainder mainly ammonium fluoride, Thermo Scientific Chemicals

Catalog No. AAL1723722
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1723722 100 g
AAL1723736 500 g
AAL172370E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1723722 Supplier Thermo Scientific Chemicals Supplier No. L1723722

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1341-49-7 | Molecular Formula: F2H5N | Molecular Weight: 57.04 g/mol

Employed in a new hydrothermal synthesis of complex fluorides NaHoF4 and NaEuF4 these interesting compounds have the fluorite structure and are important for solid-state lasers as well as for photoluminescence and magnetic characteristics

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed in a new hydrothermal synthesis of complex fluorides NaHoF4 and NaEuF4 these interesting compounds have the fluorite structure and are important for solid-state lasers as well as for photoluminescence and magnetic characteristics

Solubility
Soluble in water.

Notes
Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 1341-49-7
Molecular Formula F2H5N
Molecular Weight (g/mol) 57.04
MDL Number MFCD00012018
InChI Key KVBCYCWRDBDGBG-UHFFFAOYSA-N
Synonym Ammonium bifluoride
IUPAC Name amine dihydrofluoride
SMILES N.F.F

Specifications

Boiling Point 238°C (decomposition)
Melting Point 120°C to 122°C
pH 2 to 3
Quantity 100 g
Odor Ammonia-like
Sensitivity Hygroscopic
Density 1.5
Linear Formula (NH4)HF2
UN Number UN1727
Merck Index 14,498
Solubility Information Soluble in water.
Formula Weight 57.04
Percent Purity 95%
Assay remainder mainly ammonium fluoride
Chemical Name or Material Ammonium hydrogen fluoride
Show More Show Less
Hazard Category H301-H314-H335
Hazard Statement GHS H Statement
H300-H314-H318
Fatal if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMMONIUM HYDROGENDIFLUORIDE, SOLID
EINECSNumber 215-676-4
RTECSNumber BQ9200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only