missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hydrogen difluoride, 95%, extra pure, Thermo Scientific Chemicals

Catalog No. p-45506
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic Bottle
Plastic drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC208820010 1 kg Plastic Bottle
AC208820050 5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC208820010 Supplier Thermo Scientific Chemicals Supplier No. 208820010
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent

Chemical Identifiers

CAS 1341-49-7
Molecular Formula F2H5N
Molecular Weight (g/mol) 57.04
MDL Number MFCD00012018
InChI Key KVBCYCWRDBDGBG-UHFFFAOYSA-N
Synonym Ammonium bifluoride
IUPAC Name amine dihydrofluoride
SMILES N.F.F

Specifications

Boiling Point 230.0°C
Melting Point 125.0°C
Color White
Physical Form Flakes or Scales
Quantity 1 kg
Assay Percent Range 95%
Linear Formula (NH4)HF2
Merck Index 15, 495
Solubility Information Solubility in water: 630g/L (20°C). Other solubilities: very soluble in hot water, soluble in ethanol
Formula Weight 57.04
Percent Purity 95%
Grade Extra Pure
Packaging Plastic Bottle
Chemical Name or Material Ammonium hydrogen difluoride, tech., 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 215-676-4
RTECSNumber BQ9200000
TSCA TSCA

RUO – Research Use Only