missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hydrogen carbonate, 98%

Catalog No. p-7024237
Change view
Click to view available options
Quantity:
500 g
2500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1856636 500 g
AAA185660E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1856636 Supplier Thermo Scientific Chemicals Supplier No. A1856636
Only null left

CAS: 1066-33-7 | C3H8N2O9Zn2 | 346.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Ammonium bicarbonate is a commonly used reagent for industrial and research procedures. It acts a good buffer in lyophilization and matrix assisted laser desorption. It is also utilized for the in-gel digestion of proteins by trypsin and in the MALDI mass spectrometric analysis of proteins. It is also used to make other ammonium compounds, in food processing, and for other uses. Ammonium bicarbonate can be used to study biological buffers. It is also used in a study that demonstrated that ammonium bicarbonate salts, which can be regenerated using low-temperature waste heat, can also produce sufficient voltage for hydrogen gas generation in a microbial reverse-electrodialysis electrolysis cells. It has also been used in a study that developed a fast and sensitive method for the simultaneous determination of Sudan dyes in food samples using partial filling micellar electrokinectic chromatography-mass spectrometry.

Notes
Store away from oxidizing agents, acids and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 1066-33-7
Molecular Formula CH5NO3
Molecular Weight (g/mol) 79.06
MDL Number MFCD00012138
InChI Key ATRRKUHOCOJYRX-UHFFFAOYSA-N
Synonym ammonium bicarbonate, ammonium hydrogencarbonate, ammonium hydrogen carbonate, carbonic acid, monoammonium salt, monoammonium carbonate, ammonium acid carbonate, ammonium hydrogencarbonat, ammoniumbicarbonate, acid ammonium carbonate, ccris 7327
PubChem CID 14013
IUPAC Name carbonic acid amine
SMILES N.OC(O)=O

Specifications

Melting Point ∼60°C (decomposition)
Quantity 500 g
Beilstein 4329606
Merck Index 14,497
Solubility Information Soluble in water (220mg/ml at 20°C). Insoluble in alcohol,ethanol,benzene,and acetone.
Formula Weight 79.06
Percent Purity 98%
Odor Like Ammonia
Density 1.586
Chemical Name or Material Ammonium hydrogen carbonate
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 213-911-5
RTECSNumber BO8600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only